Learn More
Nickel Electro™ Clifton™ Stainless Steel Flat Lid
Maintain a stable bath temperature and reduce evaporation with a stainless steel lid. Nickel Electro™ Clifton™ Stainless Steel Flat Lid minimises heat loss and reduces evaporation.
£80.70 - £174.00
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
13141303
|
Nickel Electro™
LD-14 |
14 L Baths (NE1BD-18 W, NE1BD-14, NE1D-14, NE3-14 DT, NE2-5 D, NE1B-18 W, NE1B-14, NE2-14 D, NE1-14) |
£119.00
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
11766627
|
Nickel Electro™
LD-22 |
22 L Baths (NE2-28 D, NE1-28, NE2D-8/22, NE1D-22, NE3-28 DT, NE1BD-28, NE1-22, NE2-22 D, NE1B-28, NE2-9 D, NE1-9) |
£85.55
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
13131303
|
Nickel Electro™
LD-8 |
8 L Baths (NE3-8 DT, NE1D-8, NE2D-8/22, NE2-10 D, NE1-8, NE2D-4/8, NE2-8 D) |
£80.70
Each |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
11786587
|
Nickel Electro™
LD-4 |
2 L and 4 L Baths (NE3-4 DT, NE2D-4/4, NE1D-4, NE1-4, NE1D-2/2/2, NE1-4/4/4, NE2-4 D, NE1-2/2/2, NE2D-4/8, NE1D-4/4/4, NE2-2 D) |
£125.00
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
12708908
|
Nickel Electro™
LD4-14 |
14 L Baths (NE4-14 HT, NE4-14 D, NE4-14 T, NE4-14 P) |
£174.00
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
12718908
|
Nickel Electro™
LD4-22 |
22 and 28 L Baths (NE4-28 T, NE4-22 HT, NE4-28 P, NE4-22 T, NE4-22 P, NE4-28 D, NE4-22 D) |
£159.00
Each |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||||
12788898
|
Nickel Electro™
LD4-8 |
8 L Baths (NE4-8 T, NE4-8 D, NE4-8 P, NE4-8 HT) |
£114.00
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
Description
- Stainless steel
- Insulated
- Minimises heat loss
- Reduces evaporation
- Assists in maintaining bath temperature
Specifications
Stainless Steel |