Learn More
Nickel Electro™ Clifton™ Stainless Steel Flat Lid
Maintain a stable bath temperature and reduce evaporation with a stainless steel lid. Nickel Electro™ Clifton™ Stainless Steel Flat Lid minimises heat loss and reduces evaporation.
Brand:  Nickel Electro™ LD4-22
Code : 03
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12718908
  • £159.00 / Each
Estimated Shipment: 22-07-2024
to see stock.
Add to basket
Description
Description
  • Stainless steel
  • Insulated
  • Minimises heat loss
  • Reduces evaporation
  • Assists in maintaining bath temperature
Specifications
Specifications
22 and 28 L Baths (NE4-28 T, NE4-22 HT, NE4-28 P, NE4-22 T, NE4-22 P, NE4-28 D, NE4-22 D)
Stainless Steel