Learn More
Nickel Electro™ Clifton™ Stainless Steel Flat Lid
Maintain a stable bath temperature and reduce evaporation with a stainless steel lid. Nickel Electro™ Clifton™ Stainless Steel Flat Lid minimises heat loss and reduces evaporation.
Brand:  Nickel Electro™ LD4-14
Code : 03
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12708908
  • £174.00 / Each
Estimated Shipment: 14-06-2024
to see stock.
Add to basket
Description
Description
  • Stainless steel
  • Insulated
  • Minimises heat loss
  • Reduces evaporation
  • Assists in maintaining bath temperature
Specifications
Specifications
14 L Baths (NE4-14 HT, NE4-14 D, NE4-14 T, NE4-14 P)
Stainless Steel