Learn More
Nickel Electro™ Clifton™ Stainless Steel Flat Lid
Maintain a stable bath temperature and reduce evaporation with a stainless steel lid. Nickel Electro™ Clifton™ Stainless Steel Flat Lid minimises heat loss and reduces evaporation.
Brand:  Nickel Electro™ LD-4
Code : A0
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11786587
  • £125.00 / Each
Estimated Shipment: 13-06-2024
to see stock.
Add to basket
Description
Description
  • Stainless steel
  • Insulated
  • Minimises heat loss
  • Reduces evaporation
  • Assists in maintaining bath temperature
Specifications
Specifications
2 L and 4 L Baths (NE3-4 DT, NE2D-4/4, NE1D-4, NE1-4, NE1D-2/2/2, NE1-4/4/4, NE2-4 D, NE1-2/2/2, NE2D-4/8, NE1D-4/4/4, NE2-2 D)
Stainless Steel