missing translation for 'onlineSavingsMsg'
Learn More

Urocanic acid, 98%, Thermo Scientific Chemicals

£43.11 - £613.05

Chemical Identifiers

CAS 104-98-3
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00005203
InChI Key LOIYMIARKYCTBW-UPHRSURJSA-N
Synonym urocanic acid, 4-imidazoleacrylic acid, trans-urocanic acid, urocaninic acid, imidazoleacrylic acid, 5-imidazoleacrylic acid, urocanate, imidazole-4-acrylic acid, 2e-3-1h-imidazol-4-yl acrylic acid, 3-1h-imidazol-4-yl-2-propenoic acid
PubChem CID 1549103
ChEBI CHEBI:30818
IUPAC Name (Z)-3-(1H-imidazol-5-yl)prop-2-enoic acid
SMILES C1=C(NC=N1)C=CC(=O)O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10553591
View Documents Promotion Details
Thermo Scientific Acros
228960050
5 g Glass bottle
£43.11
5g
Please to purchase this item. Need a web account? Register with us today!
10206810
View Documents
Thermo Scientific Acros
228960250
25 g Glass bottle
£165.83
25g
Please to purchase this item. Need a web account? Register with us today!
10146683
View Documents
Thermo Scientific Acros
228961000
100 g Glass bottle
£613.05
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

104-98-3
138.13
LOIYMIARKYCTBW-UPHRSURJSA-N
1549103
(Z)-3-(1H-imidazol-5-yl)prop-2-enoic acid
C6H6N2O2
MFCD00005203
urocanic acid, 4-imidazoleacrylic acid, trans-urocanic acid, urocaninic acid, imidazoleacrylic acid, 5-imidazoleacrylic acid, urocanate, imidazole-4-acrylic acid, 2e-3-1h-imidazol-4-yl acrylic acid, 3-1h-imidazol-4-yl-2-propenoic acid
CHEBI:30818
C1=C(NC=N1)C=CC(=O)O

Specifications

104-98-3
White to Beige
Authentic
C6H6N2O2
25, 126
urocanic acid, 4-imidazoleacrylic acid, trans-urocanic acid, urocaninic acid, imidazoleacrylic acid, 5-imidazoleacrylic acid, urocanate, imidazole-4-acrylic acid, 2e-3-1h-imidazol-4-yl acrylic acid, 3-1h-imidazol-4-yl-2-propenoic acid
LOIYMIARKYCTBW-UPHRSURJSA-N
(Z)-3-(1H-imidazol-5-yl)prop-2-enoic acid
1549103
138.13
Powder
217.0°C to 220.0°C
0.5% max.
97.5% min. (HPLC)
MFCD00005203
10, 9691
Solubility in water: slightly soluble. Other solubilities: soluble in acetone,,insoluble in ethanol,diethyl ether
C1=C(NC=N1)C=CC(=O)O
138.13
CHEBI:30818
98%
Urocanic acid
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 203-258-4

Product Identifier
  • Urocanic acid

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.