missing translation for 'onlineSavingsMsg'
Learn More

Urocanic acid, 98%, Thermo Scientific Chemicals

Product Code. 10146683
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-98-3
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00005203
InChI Key LOIYMIARKYCTBW-UPHRSURJSA-N
Synonym urocanic acid, 4-imidazoleacrylic acid, trans-urocanic acid, urocaninic acid, imidazoleacrylic acid, 5-imidazoleacrylic acid, urocanate, imidazole-4-acrylic acid, 2e-3-1h-imidazol-4-yl acrylic acid, 3-1h-imidazol-4-yl-2-propenoic acid
PubChem CID 1549103
ChEBI CHEBI:30818
IUPAC Name (Z)-3-(1H-imidazol-5-yl)prop-2-enoic acid
SMILES C1=C(NC=N1)C=CC(=O)O

Specifications

Melting Point 217.0°C to 220.0°C
Color White to Beige
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 100 g
Beilstein 25, 126
Merck Index 10, 9691
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in acetone,,insoluble in ethanol,diethyl ether
Formula Weight 138.13
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Urocanic acid
Show More Show Less
Product Identifier
  • Urocanic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.