missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Toluidine Blue O, Certified, 65% min dye content
£22.31 - £212.06
Chemical Identifiers
CAS | 92-31-9 |
---|---|
Molecular Formula | C15H16ClN3S |
Molecular Weight (g/mol) | 305.82 |
MDL Number | MFCD00011934 |
InChI Key | HNONEKILPDHFOL-UHFFFAOYSA-M |
Synonym | Basic Blue 17,C.I. 52040 |
PubChem CID | 7083 |
IUPAC Name | (7-amino-8-methylphenothiazin-3-ylidene)-dimethylazanium;chloride |
SMILES | [Cl-].CN(C)C1=CC=C2N=C3C=C(C)C(N)=CC3=[S+]C2=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15437209
|
Thermo Scientific Chemicals
J66015.06 |
5 g |
£22.31
5g |
In Stock
Log in to see stock. |
|||||
15447209
|
Thermo Scientific Chemicals
J66015.14 |
25 g |
£41.81
25g |
In Stock
Log in to see stock. |
|||||
15457209
|
Thermo Scientific Chemicals
J66015.30 |
250 g |
£212.06
250g |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
92-31-9 | |
305.82 | |
HNONEKILPDHFOL-UHFFFAOYSA-M | |
7083 | |
[Cl-].CN(C)C1=CC=C2N=C3C=C(C)C(N)=CC3=[S+]C2=C1 |
C15H16ClN3S | |
MFCD00011934 | |
Basic Blue 17,C.I. 52040 | |
(7-amino-8-methylphenothiazin-3-ylidene)-dimethylazanium;chloride |
Specifications
92-31-9 | |
MFCD00011934 | |
Soluble in water at 1mg/ml | |
[Cl-].CN(C)C1=CC=C2N=C3C=C(C)C(N)=CC3=[S+]C2=C1 | |
305.82 | |
305.83 | |
Glass bottle | |
Toluidine Blue O |
C15H16ClN3S | |
Basic Blue 17,C.I. 52040 | |
HNONEKILPDHFOL-UHFFFAOYSA-M | |
(7-amino-8-methylphenothiazin-3-ylidene)-dimethylazanium;chloride | |
7083 | |
≥65% dye content | |
Powder |
Safety and Handling
missing translation for 'einecsNumber' : 202-146-2
missing translation for 'rtecsNumber' : SP5670000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only