missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Toluidine Blue O, Certified, 65% min dye content

Product Code. 15447209
Click to view available options
Quantity:
5 g
25 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-31-9
Molecular Formula C15H16ClN3S
Molecular Weight (g/mol) 305.82
MDL Number MFCD00011934
InChI Key HNONEKILPDHFOL-UHFFFAOYSA-M
Synonym Basic Blue 17,C.I. 52040
PubChem CID 7083
IUPAC Name (7-amino-8-methylphenothiazin-3-ylidene)-dimethylazanium;chloride
SMILES [Cl-].CN(C)C1=CC=C2N=C3C=C(C)C(N)=CC3=[S+]C2=C1

Specifications

Solubility Information Soluble in water at 1mg/ml
Formula Weight 305.83
Percent Purity ≥65% dye content
Packaging Glass bottle
Quantity 25 g
Physical Form Powder
Chemical Name or Material Toluidine Blue O

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'