missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Toluidine Blue O, Certified, 65% min dye content
Click to view available options
Quantity:
5 g
25 g
250 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Solubility Information | Soluble in water at 1mg/ml |
Formula Weight | 305.83 |
Percent Purity | ≥65% dye content |
Packaging | Glass bottle |
Quantity | 250 g |
Physical Form | Powder |
Chemical Name or Material | Toluidine Blue O |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'