missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human LRRK2, GST Tag Recombinant Protein
LRRK2 is a multidomain protein that includes a kinase domain, a GTPase domain, and a leucine-rich repeat (LRR) domain.
£479.00 - £10860.00
Specifications
Accession Number | NP_940980.2 |
---|---|
Gene Alias | LRRK2 |
KinaseFamily | LRRK Kinase Family |
Species | Human |
Recombinant | Recombinant |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15488763
|
Invitrogen™
PV4874 |
1mg |
£10860.00
1mg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15498673
|
Invitrogen™
PR8604B |
100μg |
£2290.00
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10689873
|
Invitrogen™
PV4873 |
10 μg |
£479.00
Each |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Mutations in the LRRK2 gene have been associated with Parkinson's disease.
Spécification
NP_940980.2 | |
LRRK Kinase Family | |
Recombinant | |
Baculovirus |
LRRK2 | |
Human | |
GST Tag |