missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LRRK2, GST Tag Recombinant Protein

LRRK2 is a multidomain protein that includes a kinase domain, a GTPase domain, and a leucine-rich repeat (LRR) domain.

Brand:  Invitrogen™ PV4874

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15488763

  • £10860.00 / 1mg
Estimated Shipment: 06-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Mutations in the LRRK2 gene have been associated with Parkinson's disease.

Specifications

Specifications

NP_940980.2
204.9 kDa
LRRK2
Human
GST Tag
Ser⁄Thr Kinases: TKL Group
Baculovirus
Recombinant, Catalytic
Signal Transduction
Kinase Assay
1mg
LRRK Kinase Family
Recombinant
Storage Buffer: 50 mM Tris pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Polysorbate 20, 2 mM DTT, 50% glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Dry Ice
Kinases & Inhibitors
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers