missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human LRRK2, GST Tag Recombinant Protein
LRRK2 is a multidomain protein that includes a kinase domain, a GTPase domain, and a leucine-rich repeat (LRR) domain.
Brand: Invitrogen™ PR8604B
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Mutations in the LRRK2 gene have been associated with Parkinson's disease.
Specifications
NP_940980.2 | |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Polysorbate 20, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
LRRK2 | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
204.9kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Catalytic | |
LRRK Kinase Family | |
Recombinant | |
Baculovirus |