Learn More
Fisherbrand™ 3 Tier Disc Assembly Complete
Suitable for 'End to End Mixing' tubes, these clips for tube diameters between 15 - 17mm, are ideal for 15ml Centrifuge tubes.
£172.00 - £200.00
Products 2
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
12388448
View Documents Promotion Details
Fisherbrand™
RMD012-3
12 Clips This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12378448
View Documents Promotion Details
Fisherbrand™
RMD024-3
24 Clips This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
  • Rotary Mixer Disc complete with 12 clips for tubes, supplied unassembled
  • Suitable for 'End to End Mixing' tubes
  • Clips for Tube diameters between 15 - 17mm that are suitable for 15ml Centrifuge tubes
  • The range of accessories can also be autoclaved and allow for easy loading and tube removal.
  • The accessory is easy to fit onto the RM-1 or RM-2 locating on three pins then secured in place with thumb wheel