Learn More
Fisherbrand™ 3 Tier Disc Assembly Complete
Suitable for 'End to End Mixing' tubes, these clips for tube diameters between 15 - 17mm, are ideal for 15ml Centrifuge tubes.
Brand:  Fisherbrand™ RMD024-3
 View more versions of this product
Product Code. 12378448
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Rotary Mixer Disc complete with 12 clips for tubes, supplied unassembled
  • Suitable for 'End to End Mixing' tubes
  • Clips for Tube diameters between 15 - 17 mm that are suitable for 15 mL Centrifuge tubes
  • The range of accessories can also be autoclaved and allow for easy loading and tube removal.
  • The accessory is easy to fit onto the RM-1 or RM-2 locating on three pins then secured in place with thumb wheel
Specifications
Specifications
24 Clips