Learn More
Thermo Scientific™ Barnstead™ E-Pure™ Optional Accessories
Optimize your ultrapure water purification system with Thermo Scientific™ Barnstead™ E-Pure™ Optional Accessories, including a remote dispenser.
£750.00 - £1006.77
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11486348
|
Thermo Scientific™
D8952 |
Remote Dispenser |
£1006.77
Each |
Estimated Shipment: 13-11-2024
Log in to see stock. |
|||||
12700896
|
Thermo Scientific™
D0606 |
Low Water Level Pump Protector, 240V |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Optimize your ultrapure water purification system with Thermo Scientific™ Barnstead™ E-Pure™ Optional Accessories. Increase flexibility in your lab with the remote dispenser, which delivers water up to 8' (2.4m) from the system and ships with a mounting bracket. Gain protection with the low pressure pump protector which alerts the E-Pure of an inadequate feed water condition in a pressurized feed line. The low water level pump protector alerts the system of an inadequate feed water condition when fed from a reservoir.
Specifications
Barnstead E-Pure |