missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ VibraGuard™ 7-111 Series White and Blue Nitrile Medium Weight Gloves

Minimize the risks and effects of harsh vibrations on the job.

£55.70

Specifications

Back Material Nitrile
Cuff Material Nitrile
Coating Material Nitrile
Color White, Blue
Cut Resistance Rating Level 1
View More Specs

Products 3
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
12689691
View Documents
Ansell™
7111100
10 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12609701
View Documents
Ansell™
7111090
9 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12699691
View Documents
Ansell™
7111080
8 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Industrial gloves
  • Help protect from Hand-Arm Vibration Syndrome (HAVS)
  • Half finger version for improved handling of screws, nails and small parts
  • Soft padding in the thumb, index finger and palm area enhances the shock impact protection
  • Breathable cotton back for comfort in warm work environments
  • Ansell Vibration Gel Foam padding reduces transmitted vibration to the hand
  • Nitrile coating provides snag and abrasion resistance
  • Silicone-free and launderable
  • Pressing the trigger of power tools is a lot easier with this half-finger version

Applications:

  • Handling materials with sharp or rough edges
  • Handling metal sheets and panels
  • Handling rough machined metal parts
  • Metalworking and assembly operations
Specifications

Specifications

Nitrile
Nitrile
Level 1
7-111
Hook and Loop
6 Pairs
7-111
Nitrile
White, Blue
Medium Weight Gloves
Half-Finger
Interlock Knit Cotton
Snag, puncture and abrasion resistance
Videos
SDS
Documents

Documents

Product Certifications