missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ VibraGuard™ 7-111 Series White and Blue Nitrile Medium Weight Gloves

Product Code. 12699691
Click to view available options
Size:
8
9
10
This item is not returnable. View return policy
This item is not returnable. View return policy

Minimize the risks and effects of harsh vibrations on the job.

  • Industrial gloves
  • Help protect from Hand-Arm Vibration Syndrome (HAVS)
  • Half finger version for improved handling of screws, nails and small parts
  • Soft padding in the thumb, index finger and palm area enhances the shock impact protection
  • Breathable cotton back for comfort in warm work environments
  • Ansell Vibration Gel Foam padding reduces transmitted vibration to the hand
  • Nitrile coating provides snag and abrasion resistance
  • Silicone-free and launderable
  • Pressing the trigger of power tools is a lot easier with this half-finger version

Applications:

  • Handling materials with sharp or rough edges
  • Handling metal sheets and panels
  • Handling rough machined metal parts
  • Metalworking and assembly operations
TRUSTED_SUSTAINABILITY

Specifications

Back Material Nitrile
Cuff Material Nitrile
Coating Material Nitrile
Color White, Blue
Cut Resistance Rating Level 1
Product Type Medium Weight Gloves
Size 8
Style 7-111
Finger Style Half-Finger
Glove Cuff Style Hook and Loop
Lining Interlock Knit Cotton
Quantity 6 Pairs
Resistant To Snag, puncture and abrasion resistance
Series 7-111
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'