missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP15 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | USP15 |
---|---|
Concentration | 0.44 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16892954
|
Proteintech
14354-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16882954
|
Proteintech
14354-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a member of the ubiquitin specific protease (USP) family of deubiquitinating enzymes. USP enzymes play critical roles in ubiquitin-dependent processes through polyubiquitin chain disassembly and hydrolysis of ubiquitin-substrate bonds. The encoded protein associates with the COP9 signalosome, and also plays a role in transforming growth factor beta signaling through deubiquitination of receptor-activated SMAD transcription factors. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene, and a pseudogene of this gene is located on the long arm of chromosome 2.Specifications
USP15 | |
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
Q8R5H1, Q9R085, Q9Y4E8 | |
14479, 171329, 9958 | |
USP15 Fusion Protein Ag5703 | |
Primary | |
-20°C | |
USP15 |
0.44 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Deubiquitinating enzyme 15, KIAA0529, Ubiquitin thiolesterase 15, Unph 2, UNPH4, USP15 | |
Usp15 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |