missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP15 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14354-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the ubiquitin specific protease (USP) family of deubiquitinating enzymes. USP enzymes play critical roles in ubiquitin-dependent processes through polyubiquitin chain disassembly and hydrolysis of ubiquitin-substrate bonds. The encoded protein associates with the COP9 signalosome, and also plays a role in transforming growth factor beta signaling through deubiquitination of receptor-activated SMAD transcription factors. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene, and a pseudogene of this gene is located on the long arm of chromosome 2.Specifications
USP15 | |
Polyclonal | |
Unconjugated | |
USP15 | |
Deubiquitinating enzyme 15, KIAA0529, Ubiquitin thiolesterase 15, Unph 2, UNPH4, USP15 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14479, 171329, 9958 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.44 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8R5H1, Q9R085, Q9Y4E8 | |
Usp15 | |
USP15 Fusion Protein Ag5703 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |