missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 30 Safety Storage Cabinet Q-CLASSIC-30 Left-Hinged
Safe and approved storage of flammable hazardous substances in working areas
£1619.68 - £2881.08
Specifications
Certifications/Compliance | Fire resistance 30 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
---|---|
Depth (Metric) Exterior | 620 mm |
Depth (Metric) Interior | 519 mm |
Height (Metric) Opening | 90 mm |
Material | Steel |
Alert:
For indoor use only
Product Code | Brand | Width (Metric) Exterior | Configuration | Color | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (Metric) Exterior | Configuration | Color | Price | Quantity & Availability | |||
15906256
|
asecos™
3100206133600 |
564 mm | 3 x shelf, 1 x perforated insert (380 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Gray |
£1619.68
Each |
Estimated Shipment: 12-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15966216
|
asecos™
3100206033600 |
564 mm | 3 x shelf, 1 x perforated insert (380 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Anthracite Gray, Warning Yellow |
£1619.68
Each |
Estimated Shipment: 12-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15946256
|
asecos™
3100206133608 |
564 mm | 4 x drawer (sheet steel powder-coated) | Anthracite Gray, Light Gray |
£1970.54
Each |
Estimated Shipment: 12-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15916226
|
asecos™
3100206033608 |
564 mm | 4 x drawer (sheet steel powder-coated) | Gray, Yellow |
£1970.54
Each |
Estimated Shipment: 12-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15555304
|
asecos™
3100206130088 |
564 mm | 4 x drawer, 1 x bottom collecting sump (sheet steel powder-coated) | Gray |
£2480.40
Each |
Estimated Shipment: 12-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15966256
|
asecos™
3100206133610 |
564 mm | 6 x drawer (sheet steel powder-coated) | Anthracite Gray, Light Gray |
£2250.38
Each |
Estimated Shipment: 12-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15936226
|
asecos™
3100206033610 |
564 mm | 6 x drawer (sheet steel powder-coated) | Anthracite Gray, Warning Yellow |
£2250.38
Each |
Estimated Shipment: 12-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15907026
|
asecos™
3100806133622 |
864 mm | 3 x shelf, 1 x perforated insert (380 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Gray |
£1796.17
Each |
Estimated Shipment: 12-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15361826
|
asecos™
3100806030043 |
864 mm | 3 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Gray, Yellow |
£2745.40
Each |
Estimated Shipment: 12-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15966996
|
asecos™
3100806033627 |
864 mm | 4 x drawer (sheet steel powder-coated) | Anthracite Gray, Warning Yellow |
£2438.00
Each |
Estimated Shipment: 12-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15947026
|
asecos™
3100806133629 |
864 mm | 6 x drawer (sheet steel powder-coated) | Anthracite Gray, Light Gray |
£2881.08
Each |
Estimated Shipment: 12-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15986996
|
asecos™
3100806033629 |
864 mm | 6 x drawer (sheet steel powder-coated) | Anthracite Gray, Warning Yellow |
£2881.08
Each |
Estimated Shipment: 12-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Robust construction and longevity: triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
- Easy handling with comfort: door stops in any position, self-closing in the event of fire
- No unauthorised use: door lockable with profile cylinder (integration in an existing locking system possible)
- Easy transport: integrated transport base with removable, optional base cover or adjusting aids with base cover
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
Fire resistance 30 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant | |
519 mm | |
Steel | |
Wing, Left-hinged | |
1947 mm | |
1 |
620 mm | |
90 mm | |
Safety storage cabinet | |
Safe and approved storage of flammable hazardous substances in working areas | |
1626 mm | |
Powder-coated textured |