missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 30 Safety Storage Cabinet Q-CLASSIC-30 Left-Hinged
Safe and approved storage of flammable hazardous substances in working areas
Brand: asecos™ 3100206130088
Code : NEW
Additional Details : Weight : 222.00000kg
Alert:
For indoor use only
Description
- Robust construction and longevity: triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
- Easy handling with comfort: door stops in any position, self-closing in the event of fire
- No unauthorised use: door lockable with profile cylinder (integration in an existing locking system possible)
- Easy transport: integrated transport base with removable, optional base cover or adjusting aids with base cover
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
1626 mm | |
Safety storage cabinet | |
450 mm | |
Powder-coated textured | |
564 mm | |
195.50 kg | |
Gray | |
1947 mm | |
Q30.195.056 | |
1 |
4 x drawer, 1 x bottom collecting sump (sheet steel powder-coated) | |
Safe and approved storage of flammable hazardous substances in working areas | |
519 mm | |
Steel | |
620 x 564 x 1947 mm | |
519 x 450 x 1626 mm | |
620 mm | |
Wing, Left-hinged | |
90 mm | |
Fire resistance 30 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |