missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15541-1-AP-20UL
This item is not returnable.
View return policy
Description
TRPT1 catalyzes the last step of tRNA splicing, the transfer of the splice junction 2′-phosphate from ligated tRNA to NAD to produce ADP-ribose 1′′-2′′ cyclic phosphate. It has four isoforms with MW 22-24 and 27-29 kDa.Specifications
TRPT1 | |
Polyclonal | |
Unconjugated | |
TRPT1 | |
tRNA 2 phosphotransferase 1, tRNA phosphotransferase 1, TRPT1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
107328, 293704, 83707 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q86TN4, Q8K3A2 | |
Trpt1 | |
TRPT1 Fusion Protein Ag7897 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |