missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | TRPT1 |
---|---|
Concentration | 0.15 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16894754
|
Proteintech
15541-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16884754
|
Proteintech
15541-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRPT1 catalyzes the last step of tRNA splicing, the transfer of the splice junction 2′-phosphate from ligated tRNA to NAD to produce ADP-ribose 1′′-2′′ cyclic phosphate. It has four isoforms with MW 22-24 and 27-29 kDa.Specifications
TRPT1 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q86TN4, Q8K3A2 | |
107328, 293704, 83707 | |
TRPT1 Fusion Protein Ag7897 | |
Primary | |
-20°C | |
TRPT1 |
0.15 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
tRNA 2 phosphotransferase 1, tRNA phosphotransferase 1, TRPT1 | |
Trpt1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |