Learn More
Epredia™ Lab Vision™ Tropomyosin Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Provide accurate, reproducible results with the Epredia™ Tropomyosin Ab-1, Mouse Monoclonal Antibody.
£93.20 - £468.59
Specifications
Antigen | Tropomyosin Ab-1 |
---|---|
Clone | CH1 |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12609396
|
Epredia™
MS-1256-P0 |
Concentrated | 100 μL | N/A | |||||
12629396
|
Epredia™
MS-1256-P1ABX |
Concentrated | 100 μL | N/A | |||||
12639396
|
Epredia™
MS-1256-PABX |
Concentrated | 200 μL | N/A | |||||
12619396
|
Epredia™
MS-1256-P1 |
Concentrated | 500 μL | N/A | |||||
12699386
|
Epredia™
MS-1256-P |
Concentrated | 1 mL | N/A | |||||
12669396
|
Epredia™
MS-1256-R7 |
Prediluted | 7 mL | N/A | |||||
Description
Tropomyosin is a dimeric coiled-coil protein that binds along the length of actin filaments. It is associated with the thin filaments of muscle cells and the microfilaments of nonmuscle cells. Chicken embryo fibroblasts (CEF) contain five isoforms of tropomyosin (a, b, 1, 2, and 3), identified as such by their different apparent molecular masses after separation by SDS-PAGE, but similar biochemical properties, such as resistance to heat and organic solvents, the ability to bind to F-actin filaments, and the lack of proline and tryptophan.Host Species: Mouse
Clone: CH1
Isotype: IgG1/κ
Species Reactivity: Human, Rat and Chicken. Others not known.
Epitope: Not determined
Immunogen: Tropomyosin protein purified from chicken embryo fibroblasts (CEF) and chicken leg, heart and gizzard
Positive Control: Skeletal or Cardiac muscle
Cellular Localization: Cytoplasmic Ab-1 reacts strongly with striated muscle forms of tropomyosin, including cardiac alpha-tropomyosin and skeletal alpha- and Beta-tropomyosins. ^
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting (Not verified)
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Specifications
Tropomyosin Ab-1 | |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Chicken, Human, Rat | |
IgG2a κ |
CH1 | |
Monoclonal | |
Mouse | |
Muscle Biology | |
Tropomyosin protein purified from chicken embryo fibroblasts (CEF) and chicken leg, heart, and gizzard | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.