missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Tropomyosin Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Provide accurate, reproducible results with the Epredia™ Tropomyosin Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-1256-P

 View more versions of this product

Product Code. 12699386

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Tropomyosin is a dimeric coiled-coil protein that binds along the length of actin filaments. It is associated with the thin filaments of muscle cells and the microfilaments of nonmuscle cells. Chicken embryo fibroblasts (CEF) contain five isoforms of tropomyosin (a, b, 1, 2, and 3), identified as such by their different apparent molecular masses after separation by SDS-PAGE, but similar biochemical properties, such as resistance to heat and organic solvents, the ability to bind to F-actin filaments, and the lack of proline and tryptophan.

Host Species: Mouse

Clone: CH1

Isotype: IgG1/κ

Species Reactivity: Human, Rat and Chicken. Others not known.

Epitope: Not determined

Immunogen: Tropomyosin protein purified from chicken embryo fibroblasts (CEF) and chicken leg, heart and gizzard

Positive Control: Skeletal or Cardiac muscle

Cellular Localization: Cytoplasmic Ab-1 reacts strongly with striated muscle forms of tropomyosin, including cardiac alpha-tropomyosin and skeletal alpha- and Beta-tropomyosins. ^

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not verified)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Tropomyosin Ab-1
Monoclonal
200μg/mL
Mouse
1 mL
Muscle Biology
Chicken, Human, Rat
IgG2a κ
Immunofluorescence, Immunohistochemistry (Paraffin)
CH1
Unconjugated
Tropomyosin protein purified from chicken embryo fibroblasts (CEF) and chicken leg, heart, and gizzard
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.