Learn More
Tocris Bioscience™ ICI 174,864
δ selective peptide antagonist
£121.73
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18036062
|
Tocris Bioscience™
0820/1 |
1 mg |
£121.73
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Selective δ opioid antagonist. Exhibits partial agonist in vitro activity at δ receptors at high concentrations.Specifications
691.87 |