Learn More
Tocris Bioscience™ ICI 174,864
δ selective peptide antagonist
Brand:  Tocris Bioscience™ 0820/1
Additional Details : Weight : 0.00970kg
 View more versions of this product
Product Code. 18036062
  • £121.73 / 1mg
Estimated Shipment: 18-06-2024
to see stock.
Description
Description
ICI 174,864 is a selective δ opioid antagonist. Exhibits partial agonist in vitro activity at δ receptors at high concentrations.
Specifications
Specifications
0.95
Store at -20°C
691.87