missing translation for 'onlineSavingsMsg'
Learn More

TMEM120A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen TMEM120A
Concentration 0.21 mg/mL
Applications Immunocytochemistry, Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16897344
View Documents
Proteintech
17455-1-AP-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
16887344
View Documents
Proteintech
17455-1-AP-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TMEM120A is a transmembrane protein. The function of TMEM120A has not been widely studied, and is yet to be fully elucidated.
Specifications

Specifications

TMEM120A
Immunocytochemistry, Western Blot, Immunofluorescence
Unconjugated
Rabbit
Rat, Mouse, Human
Q5HZE2, Q8C1E7, Q9BXJ8
215210, 288591, 83862
TMEM120A Fusion Protein Ag10849
Primary
-20°C
TMEM120A
0.21 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
NET29, TMEM120A, TMPIT, transmembrane protein 120A
Tmem120a
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications