missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM120A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17455-1-AP-150UL
This item is not returnable.
View return policy
Description
TMEM120A is a transmembrane protein. The function of TMEM120A has not been widely studied, and is yet to be fully elucidated.Specifications
TMEM120A | |
Polyclonal | |
Unconjugated | |
TMEM120A | |
NET29, TMEM120A, TMPIT, transmembrane protein 120A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
215210, 288591, 83862 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5HZE2, Q8C1E7, Q9BXJ8 | |
Tmem120a | |
TMEM120A Fusion Protein Ag10849 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |