missing translation for 'onlineSavingsMsg'
Learn More

TAC1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen TAC1
Concentration 0.33 mg/mL
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16832234
View Documents
Proteintech
13839-1-AP-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
16822234
View Documents
Proteintech
13839-1-AP-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This antibody is specific to TAC1.

This gene encodes four products of the tachykinin peptide hormone family, substance P and neurokinin A, as well as the related peptides, neuropeptide K and neuropeptide gamma. These hormones are thought to function as neurotransmitters which interact with nerve receptors and smooth muscle cells. They are known to induce behavioral responses and function as vasodilators and secretagogues. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

TAC1
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot
Unconjugated
Rabbit
Human, Mouse
P20366, P41539
21333, 6863
TAC1 Fusion Protein Ag4790
Primary
-20°C
TAC1
0.33 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Hs.2563, Neuromedin L, NK2, NKA, NKNA, PPT, Protachykinin 1, Substance K Neuropeptide K, Substance P, SubstanceP, TAC1, TAC2, tachykinin, precursor 1
TAC1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications