missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAC1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13839-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is specific to TAC1.
This gene encodes four products of the tachykinin peptide hormone family, substance P and neurokinin A, as well as the related peptides, neuropeptide K and neuropeptide gamma. These hormones are thought to function as neurotransmitters which interact with nerve receptors and smooth muscle cells. They are known to induce behavioral responses and function as vasodilators and secretagogues. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
TAC1 | |
Polyclonal | |
Unconjugated | |
TAC1 | |
Hs.2563, Neuromedin L, NK2, NKA, NKNA, PPT, Protachykinin 1, Substance K Neuropeptide K, Substance P, SubstanceP, TAC1, TAC2, tachykinin, precursor 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21333, 6863 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P20366, P41539 | |
TAC1 | |
TAC1 Fusion Protein Ag4790 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |