Learn More
Superfos™ SuperLift™ Polypropylene Standard with Centering Lid
Manufactured from polypropylene. Superfos™ SuperLift™ Polypropylene Standard with Centering Lid includes tamper evidence while the protective collar safeguards the lid to prevent container ovalisation.
£0.26
Specifications
Product Type | Standard with Centering Lid |
---|---|
Material | Polypropylene |
Autoclavable | Autoclavable |
Certifications/Compliance | European Union Commission Regulation No. 10/2011/EC amendments |
Includes | Standard Lid with Centering Pieces |
Description
- Manufactured from polypropylene
- Includes tamper evidence while the protective collar safeguards the lid and prevents container ovalisation.
- Very good heat resistance
- Packaging is suitable for hot filling.
- Notably, plastic packaging become flexible when subjected to hot filling temperatures and care must be taken in relation to stacking immediately after hot filling.
- Freezing conditions require special resins available on request.
Specifications
Standard with Centering Lid | |
Autoclavable | |
Standard Lid with Centering Pieces |
Polypropylene | |
European Union Commission Regulation No. 10/2011/EC amendments | |
1 Ea. |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.