Learn More
Superfos™ SuperLift™ Polypropylene Standard with Centering Lid
Manufactured from polypropylene. Superfos™ SuperLift™ Polypropylene Standard with Centering Lid includes tamper evidence while the protective collar safeguards the lid to prevent container ovalisation.
Brand:  Superfos™ 1671
Encompass
 View more versions of this product
Product Code. 10228014
  • £0.26 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • Manufactured from polypropylene
  • Includes tamper evidence while the protective collar safeguards the lid and prevents container ovalisation.
  • Very good heat resistance
  • Packaging is suitable for hot filling.
  • Notably, plastic packaging become flexible when subjected to hot filling temperatures and care must be taken in relation to stacking immediately after hot filling.
  • Freezing conditions require special resins available on request.
Specifications
Specifications
Standard with Centering Lid
Autoclavable
223.7 mm
12.5 mm
1 Ea.
Polypropylene
European Union Commission Regulation No. 10/2011/EC amendments
3.5 and 5.8L Bucket
Standard Lid with Centering Pieces