missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ SteriWare™ PharmaScoops™ PP (White)
BIOPHARMA

Our single use, White SteriWare™ PharmaScoops™ PP give additional chemical resistance over conventional plastic scoops and are available from stock in a huge range of sizes.

Brand:  Sampling Systems™ 8045P-2500

 View more versions of this product

Product Code. 17504586

  • £69.10 / Pack of 15
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The Sampling Systems range of sterile sampling scoops are used by companies all over the world.
  • They are available in a wide range of materials and colors.
  • PP gives excellent chemical resistance.
  • Our sterile, disposable Scoops (spades) are manufactured in a class 100,000 cleanroom to ensure cleanliness.
  • They are all individually bagged and so can be used straight from the bag without the need for costly cleaning.
  • They are available from stock in a huge range of sizes from 30 mL (1 oz.) up to 2500 mL (88 oz.).
  • Standard stock sizes are 30, 60, 125, 250, 500, 1000 and 2500 mL.
  • PharmaScoops™ are ideal for both sampling and dispensing powders and granules.
  • The Sampling Systems range of disposable PharmaScoops™ are the ultimate in quality and affordability.
  • Made from FDA and EU conforming materials.
  • BSE/TSE Free.
  • Available in red, blue or white.
  • Full Batch Traceability.
  • The scoops are also available as gamma irradiated sterile in conformance to (SAL 10-6).
  • They are available from stock for immediate delivery worldwide.
Specifications

Specifications

Polypropylene
Single-use
White
2500 mL
Non-sterile
Individually Bagged
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.