Learn More
OHAUS™ Specialty Water Bath Clamp
OHAUS™ Specialty Water Bath Clamp designed to securely grip and position laboratory glassware.
£35.50 - £47.10
Specifications
Type | Specialty Water Bath Clamp |
---|---|
Material | Nickel-plated |
For Use With (Equipment) | Water Bath |
Description
- Choose from a variety of specialized LabJaws clamps with adjustable features to provide an exact fit for lab ware such as thermometers, thermocouples, columns, funnels, and round bottom flasks.
- Versatile multi-purpose LabJaws clamps securely hold glassware and irregularly-shaped objects.
- Nickel-plated zinc clamps offer high-tensile strength at an economical price.
Specifications
Specialty Water Bath Clamp | |
Nickel-plated | |
Water Bath |