missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Fluoride, ≥99%, Reagent Grade, Honeywell™
£22.06 - £108.87
Chemical Identifiers
CAS | 7681-49-4 |
---|---|
Molecular Formula | FNa |
Molecular Weight (g/mol) | 41.99 |
MDL Number | MFCD00003524 |
InChI Key | PUZPDOWCWNUUKD-UHFFFAOYSA-M |
Synonym | sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril |
PubChem CID | 5235 |
ChEBI | CHEBI:28741 |
IUPAC Name | sodium;fluoride |
SMILES | [F-].[Na+] |
Description
Chemical Identifiers
7681-49-4 | |
41.99 | |
PUZPDOWCWNUUKD-UHFFFAOYSA-M | |
5235 | |
sodium;fluoride |
FNa | |
MFCD00003524 | |
sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril | |
CHEBI:28741 | |
[F-].[Na+] |
Specifications
1704°C (1013hPa) | |
7681-49-4 | |
8.0 to 10.0 (at 20°C) | |
FNa | |
MFCD00003524 | |
sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril | |
[F-].[Na+] | |
41.99 | |
CHEBI:28741 | |
≥99% | |
1.4mmHg (0°C) | |
Sodium fluoride |
990°C | |
Colorless | |
Solid | |
NaF | |
UN1690 | |
PUZPDOWCWNUUKD-UHFFFAOYSA-M | |
sodium;fluoride | |
5235 | |
41.99g/mol | |
Odorless | |
2.7g/cu.cm at 20°C |
Safety and Handling
Missing Phrase - N15.00950417-P305 + P351 + P338
missing translation for 'dotInformation' : 6.1/PGIII
missing translation for 'rtecsNumber' : 231-667-8