missing translation for 'onlineSavingsMsg'
Learn More

Sodium Fluoride, ≥99%, Reagent Grade, Honeywell™

Product Code. 15688070
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7681-49-4
Molecular Formula FNa
Molecular Weight (g/mol) 41.99
MDL Number MFCD00003524
InChI Key PUZPDOWCWNUUKD-UHFFFAOYSA-M
Synonym sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril
PubChem CID 5235
ChEBI CHEBI:28741
IUPAC Name sodium;fluoride
SMILES [F-].[Na+]

Specifications

Boiling Point 1704°C (1013hPa)
Melting Point 990°C
Color Colorless
pH 8.0 to 10.0 (at 20°C)
Physical Form Solid
Quantity 500 g
Linear Formula NaF
UN Number UN1690
Formula Weight 41.99g/mol
Percent Purity ≥99%
Odor Odorless
Packaging Poly bottle
Vapor Pressure 1.4mmHg (0°C)
Density 2.7g/cu.cm at 20°C
Chemical Name or Material Sodium fluoride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'