missing translation for 'onlineSavingsMsg'
Learn More

Ansell Edmont™ Sensilite™ 48-121 Series Black Polyurethane Light Weight Gloves

An economical hand protection solution offering good grip, dexterity and comfort. The polyester liner offers good hand protection in lightweight applications.

£18.13

Specifications

Back Material Polyurethane
Certifications/Compliance CE Mark, EN 388 : 3121
Cuff Material Polyurethane
Coating Material Polyurethane
Product Type Lightweight Gloves
View More Specs

Products 6
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
12630942
View Documents
Ansell Edmont™
48121070
7 N/A
N/A
12600942
View Documents
Ansell Edmont™
48121100
10 N/A
N/A
12640942
View Documents
Ansell Edmont™
48121080
8 N/A
N/A
12610942
View Documents
Ansell Edmont™
48121110
11 N/A
N/A
12620942
View Documents
Ansell Edmont™
48121060
6 N/A
N/A
12650942
View Documents
Ansell Edmont™
48121090
9 N/A
N/A
Description

Description

  • For assembly of small pieces such as small bearings, joints and springs
  • Polyester liner
  • Knitted construction
  • Polyurethane coating
  • Palm-coated finish
  • Knitwrist cuff
Specifications

Specifications

Polyurethane
Polyurethane
Lightweight Gloves
Knitwrist
Coated-palm
Tears and abrasions
CE Mark, EN 388 : 3121
Polyurethane
48-121
Polyester
144 Pairs
48-121
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.