missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences Tygon™ E-1000 Tubing

Flexible, food and beverage dispensing Tygon™ E-1000 Tubing.

Brand:  Saint Gobain Life Sciences ADK00003

Code : 02

Additional Details : Weight : 1.10000kg

 View more versions of this product

Product Code. 13133603

  • £89.22 / 15m
Estimated Shipment: 05-06-2024
to see stock.
Add to basket

Description

Description

Soft and flexible, Tygon™ E-1000 tubing delivers superior performance in a formulation that contains non-DEHP [Bis (2-ethylhexyl) phthalate] plasticizers. Tygon™ E-1000 non-DEHP tubing has been tested rigorously to meet stringent standards for low temperature and corrosive chemical resistance.

  • Ultra-smooth inner bore reduces potential for particle entrapment
  • Minimal extractable help maintain fluid integrity
  • Excellent fluid flow characteristics
  • Complete inventory of standard sizes available, including metric sizes

Typical Applications

  • Food and beverage dispensing
Specifications

Specifications

Tubing
1.6 mm
3/16 in.
50 ft.
1/16 in.
26 psi
1/16 in.
Tygon E-1000
4.8 mm
15 m
1.6 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers