missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Cys-Protein-G Protein
A cDNA sequence encoding the Cys-Protein-G was constructed and used to recombinantly synthesize the protein.
£99.15 - £1655.00
Specifications
Name | Cys-Protein-G Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15947728
|
enQuireBio™
QP11587-1MG |
1 mg |
£99.15
1mg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15937728
|
enQuireBio™
QP11587-10MG |
10 mg |
£283.00
10mg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15927728
|
enQuireBio™
QP11587-100MG |
100 mg |
£1655.00
100mg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Cys-Protein-G Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Greater than 95.0% as determined by SDS-PAGE and HPLC analyses. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Lyophilized without any additives. |