missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Cys-Protein-G Protein
A cDNA sequence encoding the Cys-Protein-G was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11587-10mg
Additional Details : Weight : 0.01000kg
Specifications
Cys-Protein-G Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Lyophilized without any additives. |
10 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Greater than 95.0% as determined by SDS-PAGE and HPLC analyses. |