missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Bromide, Certified AR for Analysis, Fisher Chemical™
£62.35 - £217.00
Chemical Identifiers
CAS | 7758-02-3 |
---|---|
Molecular Formula | BrK |
Molecular Weight (g/mol) | 119.00 |
MDL Number | MFCD00011358 |
InChI Key | IOLCXVTUBQKXJR-UHFFFAOYSA-M |
Synonym | potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095 |
PubChem CID | 253877 |
ChEBI | CHEBI:32030 |
SMILES | [K+].[Br-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10705151
|
Fisher Chemical
P/4000/50 |
250 g |
£62.35
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10385950
|
Fisher Chemical
P/4000/53 |
500 g |
£90.80
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10676072
|
Fisher Chemical
P/4000/60 |
1 kg |
£217.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
7758-02-3 | |
119.00 | |
IOLCXVTUBQKXJR-UHFFFAOYSA-M | |
253877 | |
[K+].[Br-] |
BrK | |
MFCD00011358 | |
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095 | |
CHEBI:32030 |
Specifications
1435°C | |
7758-02-3 | |
5 to 8.8 | |
BrK | |
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095 | |
[K+].[Br-] | |
119.00 | |
CHEBI:32030 | |
Plastic powder jar |
734°C | |
White | |
Solid | |
MFCD00011358 | |
IOLCXVTUBQKXJR-UHFFFAOYSA-M | |
potassium bromide | |
253877 | |
119g/mol | |
<0.01mmHg at 20°C |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Potassium bromide
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.