missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bromide, Certified AR for Analysis, Fisher Chemical™

Product Code. 10676072
Click to view available options
Quantity:
250 g
500 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7758-02-3
Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
PubChem CID 253877
ChEBI CHEBI:32030
SMILES [K+].[Br-]

Specifications

Boiling Point 1435°C
Melting Point 734°C
Color White
pH 5 to 8.8
Physical Form Solid
Quantity 1 kg
IUPAC Name potassium bromide
Formula Weight 119g/mol
Packaging Plastic powder jar
Vapor Pressure <0.01mmHg at 20°C
compliance-icons
Product Identifier
  • Potassium bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'