missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Polypropylene Male Compression to NPT(M) Adapter Elbows

These Male Compression Pipe Adapter Fittings work well with rigid tubing, including tubing made from fluoropolymers, other rigid plastics and metals.

Brand:  Masterflex™ 06386-20

 View more versions of this product

Product Code. 11701204

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Male pipe adapter elbow connector compression fitting
  • Polypropylene construction
  • Available in packs of ten
  • Very good chemical resistance
  • Rated to 220psi maximum at 25°C 
  • Usable from 0° to 100°C 
  • Compression fitting with nut that compresses rigid tubing against fitting body
  • No leaks, flow restrictions or costly downtime
Specifications

Specifications

Compression Fitting
1/4 tubing O.D. x 1/4 in. NPT(M)
10 Pack
0.25 in. NPT(M)
220 psi
Polypropylene
0°C
9/32 in.; 1 x 1-11/32 in.
Male pipe adapter elbow connector; Ethylene Oxide sterilization,
100°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers