Learn More
Kimberly-Clark™ KleenGuard™ A71 Chemical Spray Protection Coveralls
Coveralls create a spray-tight and liquid-tight barrier against harmful substances when working with hazardous chemicals.
£292.00
Specifications
Product Type General Purpose Coveralls
Material Spunbond Polypropylene
Color Yellow
Disposable Reusable
Waist Style Elastic
View More Specs
Products 2
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
12631396
View Documents Promotion Details
Kimberly-Clark™
96770
Large
£292.00
Pack of 10
Estimated Shipment: 21-03-2025
to see stock.
Add to basket
12621396
View Documents
Kimberly-Clark™
96760
Medium
£292.00
Pack of 10
Estimated Shipment: 11-06-2025
to see stock.
Add to basket
Description
Description
  • Approved as Complex Design (Category 3) equipment - offering protection to the levels specified for Type 3, 4 and 5 by CEN
  • Highly visible yellow color - for improved worker safety
  • Ideal for working with hazardous chemicals - protecting against liquid, aerosol and dry chemical splashes, sprays and dusts in low-pressure industrial cleaning and maintenance areas
  • Single-use protective clothing to improve safety and comfort in high-risk areas - create a spray and liquid tight barrier against harmful chemical splashes, liquids or dust particles
  • Suitable for use as protection against Infective Agents and Biohazards
  • Suitable for use as protection against solid particles and particulate radioactive contamination
  • Treated to be antistatic
  • EN 1073-2 approved for radioactive dust protection
  • Anti-static EN 1149-1 for critical areas
Specifications
Specifications
General Purpose Coveralls
Yellow
Elastic
Elastic
Elastic
Stitched
Type 3, 4 and 5
Yes
Hazardous Chemicals
Spunbond Polypropylene
Reusable
Elastic
Hooded
Zipper
Non-sterile
Perforation, Flame, Abrasion, Tears, Particle, Liquid
None
Full Body
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Kimberly-Clark™ KleenGuard™ A71 Chemical Spray Protection Coveralls