missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kimberly-Clark™ KleenGuard™ A71 Chemical Spray Protection Coveralls
Click to view available options
Size:
Medium
Large
Description
- Approved as Complex Design (Category 3) equipment - offering protection to the levels specified for Type 3, 4 and 5 by CEN
- Highly visible yellow color - for improved worker safety
- Ideal for working with hazardous chemicals - protecting against liquid, aerosol and dry chemical splashes, sprays and dusts in low-pressure industrial cleaning and maintenance areas
- Single-use protective clothing to improve safety and comfort in high-risk areas - create a spray and liquid tight barrier against harmful chemical splashes, liquids or dust particles
- Suitable for use as protection against Infective Agents and Biohazards
- Suitable for use as protection against solid particles and particulate radioactive contamination
- Treated to be antistatic
- EN 1073-2 approved for radioactive dust protection
- Anti-static EN 1149-1 for critical areas
Specifications
Specifications
Product Type | General Purpose Coveralls |
Size | Medium |
Material | Spunbond Polypropylene |
Color | Yellow |
Disposable | Reusable |
Waist Style | Elastic |
Ankle Style | Elastic |
Cuff Style | Elastic |
Neck Style | Hooded |
Hood Type | Elastic |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction