missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLEKHA9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | PLEKHA9 |
---|---|
Concentration | 0.22 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16897484
|
Proteintech
17568-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16887484
|
Proteintech
17568-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The function of PLEKHA9 remains unknown.Specifications
PLEKHA9 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
D3ZY60, O95397, Q80W71 | |
231999, 500132, 51054 | |
PLEKHA9 Fusion Protein Ag11772 | |
Primary | |
-20°C | |
PLEKHA8P1 |
0.22 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
PLEKHA9, Protein PLEKHA9 | |
Plekha8, PLEKHA8P1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |