missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLEKHA9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17568-1-AP-20UL
This item is not returnable.
View return policy
Description
The function of PLEKHA9 remains unknown.Specifications
PLEKHA9 | |
Polyclonal | |
Unconjugated | |
PLEKHA8P1 | |
PLEKHA9, Protein PLEKHA9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
231999, 500132, 51054 | |
-20°C | |
Liquid |
Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
D3ZY60, O95397, Q80W71 | |
Plekha8, PLEKHA8P1 | |
PLEKHA9 Fusion Protein Ag11772 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |