Learn More
Placental Alk. Phosphatase (PLAP) Ab-1 Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ Placental Alk. Phosphatase (PLAP) Ab-1, Mouse Monoclonal Antibody.
£104.04 - £403.00
Specifications
Antigen | Placental Alk. Phosphatase (PLAP) Ab-1 |
---|---|
Clone | 8B6.18 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Host Species | Mouse |
Product Code | Brand | Conjugate | Concentration | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Conjugate | Concentration | Quantity | Price | Quantity & Availability | |||
12670547
|
Epredia
MS-208-P1 |
Unconjugated | 200μg/mL | 500 μL |
£307.00
500µL |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12660547
|
Epredia
MS-208-P1ABX |
Unconjugated | 1mg/mL | 100 μL |
£345.00
100µL |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12650547
|
Epredia
MS-208-P0 |
Unconjugated | 200μg/mL | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12620547
|
Epredia
MS-208-B0 |
Biotin | 200μg/mL | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12630547
|
Epredia
MS-208-B1 |
Biotin | 200μg/mL | 500 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12680547
|
Epredia
MS-208-PABX |
Unconjugated | 1mg/mL | 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Host Species: Mouse
Clone: 8B6.18
Isotype: IgG2a/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: HEp-2 cells expressing Placental Alkaline Phosphatase (PLAP)
Molecular Weight: 60-70kDa
Positive Control: Placenta
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- ELISA
Note:
USA: RUO; Int'l: RUO
Specifications
Placental Alk. Phosphatase (PLAP) Ab-1 | |
Immunohistochemistry (Paraffin) | |
Mouse | |
Cancer and Tumor Biology | |
HEp-2 cells expressing Placental Alkaline Phosphatase (PLAP) | |
Primary |
8B6.18 | |
Monoclonal | |
RUO | |
Human | |
IgG2a κ |