Learn More
Placental Alk. Phosphatase (PLAP) Ab-1 Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ Placental Alk. Phosphatase (PLAP) Ab-1, Mouse Monoclonal Antibody.
Brand: Epredia MS-208-B0
Code : LD
Additional Details : Weight : 1.00000kg
Description
Host Species: Mouse
Clone: 8B6.18
Isotype: IgG2a/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: HEp-2 cells expressing Placental Alkaline Phosphatase (PLAP)
Molecular Weight: 60-70kDa
Positive Control: Placenta
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- ELISA
Note:
USA: RUO; Int'l: RUO
Specifications
Placental Alk. Phosphatase (PLAP) Ab-1 | |
Monoclonal | |
200μg/mL | |
Mouse | |
Placenta | |
RUO | |
Primary | |
Concentrated |
Immunohistochemistry (Paraffin) | |
8B6.18 | |
Biotin | |
HEp-2 cells expressing Placental Alkaline Phosphatase (PLAP) | |
100 μL | |
Cancer and Tumor Biology | |
Human | |
IgG2a κ |